MilliporeSigma
HomeReaction Design & OptimizationTetrakis(dimethylamino)ethylene (TDAE)
Tetrakis(dimethylamino)ethylene (TDAE)
Mark Redlich
In 2001, Professor William Dolbier, Jr., at the University of Florida reported1 an approach to nucleophilic trifluoromethylation based on the generation of a trifluoromethyl anion using CF3I in the presence of a powerful twoelectron reductant, tetrakis(dimethylamino)ethylene (TDAE) (Scheme 1).
Scheme 1
Scheme 1
Over the ensuing years, Dolbier’s group has developed numerous reactions featuring this reagent system, including the trifluoromethylation of aldehydes and ketones,1 cyclic sulfates,2 disulfides and diselenides,3 and imines1 (Scheme 2). In many cases, the TDAE/CF3I method provided results that complement other nucleophilic trifluoromethylation reagents. Recently, Dolbier has expanded the use of his system to perfluoroalkylation,4 and was able to achieve similar results as with CF3I; however, when the perfluorobutyl system was used, yields tended to decrease noticeably.
Scheme 2
Scheme 2
TDAE has a reducing power comparable to zinc metal and as such has also been used as a reducing agent in organic synthesis. Recently, Vanelle and coworkers have exploited this reductive property to obtain epoxides from aldehydes and 2-(dibromomethyl)quinoxaline (Scheme 3),5 as well as α-chloroketones from aldehydes and 2-(trichloromethyl)-substituted azaheterocycles (Scheme 4).6 The Nishiyama group at Kansai University in Osaka has also used TDAE to obtain 1,2,3,4-tetrahydronaphthalenes from 1,2-bis(bromomethyl)arenes and olefins,7 and 1,4-diketones or diesters through reductive coupling of α-bromoketones or esters (Scheme 5).8
Scheme 3
Scheme 3
Scheme 4
Scheme 4
Scheme 5
Scheme 5
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Aït-Mohand S, Takechi N, Médebielle M, Dolbier WR. 2001. Nucleophilic Trifluoromethylation Using Trifluoromethyl Iodide. A New and Simple Alternative for the Trifluoromethylation of Aldehydes and Ketones. Org. Lett.. 3(26):4271-4273. https://doi.org/10.1021/ol016933x
2.
Takechi N, Ait-Mohand S, Medebielle M, Dolbier WR. 2002. Novel Nucleophilic Trifluoromethylation of Vicinal Diol Cyclic Sulfates. Org. Lett.. 4(26):4671-4672. https://doi.org/10.1021/ol0270374
3.
Pooput C, Medebielle M, Dolbier WR. 2004. A New and Efficient Method for the Synthesis of Trifluoromethylthio- and Selenoethers. Org. Lett.. 6(2):301-303. https://doi.org/10.1021/ol036303q
4.
Pooput C, Dolbier, WR, Médebielle M. 2006. Nucleophilic Perfluoroalkylation of Aldehydes, Ketones, Imines, Disulfides, and Diselenides. J. Org. Chem.. 71(9):3564-3568. https://doi.org/10.1021/jo060250j
5.
Montana M, Terme T, Vanelle P. 2005. Original synthesis of oxiranes via TDAE methodology: reaction of 2,2-dibromomethylquinoxaline with aromatic aldehydes. Tetrahedron Letters. 46(48):8373-8376. https://doi.org/10.1016/j.tetlet.2005.09.152
6.
Montana M, Terme T, Vanelle P. 2006. Original synthesis of ?-chloroketones in azaheterocyclic series using TDAE approach. Tetrahedron Letters. 47(37):6573-6576. https://doi.org/10.1016/j.tetlet.2006.07.030
7.
Nishiyama Y, Kawabata H, Kobayashi A, Nishino T, Sonoda N. 2005. Reductive debromination of 1,2-bis(bromomethyl)arenes using tetrakis(dimethylamino)ethylene (TDAE). Tetrahedron Letters. 46(5):867-869. https://doi.org/10.1016/j.tetlet.2004.11.114
8.
Nishiyama Y, Kobayashi A. 2006. Synthesis of 1,4-diketones: reaction of ?-bromo ketones with tetrakis(dimethylamino)ethylene (TDAE). Tetrahedron Letters. 47(31):5565-5567. https://doi.org/10.1016/j.tetlet.2006.05.141