MilliporeSigma
HomeReaction Design & OptimizationBis(2-methoxyethyl)aminosulfur Trifluoride (Deoxo-Fluor®)
Bis(2-methoxyethyl)aminosulfur Trifluoride (Deoxo-Fluor®)
Originally reported by Lal and coworkers in 1999,1 bis(2-methoxyethyl)aminosulfur trifluoride (Deoxo-Fluor®) has shown remarkable utility in organic synthesis as a thermally stable alternative to (diethylamino)sulfur trifluoride (DAST). Deoxo-Fluor® can easily convert alcohols to alkyl fluorides, aldehydes and ketones to gem-difluorides, and carboxylic acids to acid fluorides or trifluoromethyl derivatives (Scheme 1).2
Trifluoromethyl derivatives
Scheme 1.
A recent report showed the use of Deoxo-Fluor® in the synthesis of a 2-azabicyclo[2.1.1]hexane analogue of 4-fluoroproline 
(Scheme 2).3 Although the reaction generally proceeds with inversion at the chiral carbon, the configuration is retained in this constrained substrate due to neighboring-group participation of the amide group.
Hexane analogue of 4-fluoroproline
Scheme 2.
Deoxo-Fluor® can be used to effect a wide array of additional transformations. One recent example highlights its use in the syntheses of a series of chiral C2 bis-oxazoline ligands (Scheme 3).4 The chemoselectivity of Deoxo-Fluor® was superior to other reagents, including DAST, and was tolerant of steric and electronic variations within the ligands. Furthermore, the Deoxo-Fluor® protocol also allowed the majority of the ligands to be purified without requiring chromatography.
Chiral C2 bis-oxazoline ligands
Scheme 3.
Gunda Georg at the University of Kansas has reported the use of Deoxo-Fluor® in a one-flask protocol to convert acids to amides and peptides or Weinreb amides (Scheme 4).5 The reaction proceeded under mild conditions and provided the desired products in high yields with facile purification.
Weinreb amides
Scheme 4.
The Kangani group has devoted several publications to one-pot transformations achievable with Deoxo-Fluor®.6 Generally, the reactions begin with the conversion of a carboxylic acid to an acid fluoride, which is then reacted with various nucleophiles to produce oxazolines, aldehydes, ketones, benzoxazoles, oxadiazoles, acyl azides or nitriles (Scheme 5).
acyl azides or nitriles
Scheme 5.
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
(a) Lal, G. S. et al. J. Org. Chem. 1999, 64, 7048. (b) Lal, G. S. et al. Chem. Commun. 1999, 215. (LGSeaJOC167(LGSeaCC12.
2.
Shreeve JM, Singh RP. 2002. Recent Advances in Nucleophilic Fluorination Reactions of Organic Compounds­ Using Deoxofluor and DAST. Synthesis.(17):2561-2578. https://doi.org/10.1055/s-2002-35626
3.
Jenkins CL, Lin G, Duo J, Rapolu D, Guzei IA, Raines RT, Krow GR. 2004. Substituted 2-Azabicyclo[2.1.1]hexanes as Constrained Proline Analogues:  Implications for Collagen Stability. J. Org. Chem.. 69(25):8565-8573. https://doi.org/10.1021/jo049242y
4.
Albano VG, Bandini M, Monari M, Marcucci E, Piccinelli F, Umani-Ronchi A. 2006. Synthesis and Crystallographic Characterization of Chiral Bis-oxazoline-amides. Fine-Tunable Ligands for Pd-Catalyzed Asymmetric Alkylations. J. Org. Chem.. 71(17):6451-6458. https://doi.org/10.1021/jo060767v
5.
White JM, Tunoori AR, Turunen BJ, Georg GI. 2004. [Bis(2-methoxyethyl)amino]sulfur Trifluoride, the Deoxo-Fluor Reagent:  Application toward One-Flask Transformations of Carboxylic Acids to Amides. J. Org. Chem.. 69(7):2573-2576. https://doi.org/10.1021/jo035658k
6.
Kangani CO, Kelley DE. 2005. One pot direct synthesis of amides or oxazolines from carboxylic acids using Deoxo-Fluor reagent. Tetrahedron Letters. 46(51):8917-8920. https://doi.org/10.1016/j.tetlet.2005.10.068