MilliporeSigma
HomeDalPhos Ligands
DalPhos Ligands
Professor Mark Stradiotto and coworkers at Dalhousie University have developed air-stable Dalphos, which contains the bulky di(1-adamantyl)phosphino [P(1-Ad)2] fragment. These chelating N,P ligands are useful for previously unprecedented Pd-catalyzed C-N and C-C bond formation. Both Me-DalPhos (Cat. No. 751413) and Mor-DalPhos (Cat. No. 751618) allow for Pd-catalyzed ammonia cross-coupling. In particular, the more reactive Mor-Dalphos improved the scope and utility of ammonia coupling at room temperature.
Benefits of DalPhos Ligands:
  • Monoarylation of ammonia, hydrazine and acetone
  • Air-stable
  • Good functional group tolerance
  • Mild conditions
Me-DalPhos featuring an ortho-dimethylamino group has been shown to be a valuable ligand for the Pd-catalyzed cross-coupling of aryl chlorides and amines (including ammonia). The highly efficient Mor-Dalphos, where the dimethylamino group is replaced by morpholine, easily enables the ammonia coupling of a range of aryl chlorides and aryl tosylates.
Cross-Coupling of Ammonia
Mor-DalPhos can be used for the first Pd-catalyzed cross-coupling of aryl chlorides and tosylates with hydrazine. The Stradiotto group also explored the scope of the reaction of aryl hydrazines with benzaldehydes to afford the corresponding aryl hydrazones.
Cross-Coupling of Hydrazine
The direct arylation of acetone with a range of aryl halides and tosylates has been reported with good monoarylation selectivity. The use of Mor-DalPhos has a key impact on the rate and selectivity of this reaction.
Cross-Coupling of Acetone
The use of Mor-DalPhos for the gold-catalyzed hydroamination of internal aryl alkynes with dialkylamines has been studied. This gold precatalyst featuring a P,N-ligand effects the stereoselective addition of a variety of functionalized dialkylamines to unsymmetrical internal arylacetylenes.
Gold-Catalyzed Hydroamination
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Lundgren R, Peters B, Alsabeh P, Stradiotto M. 2010. A P,N-Ligand for Palladium-Catalyzed Ammonia Arylation: Coupling of Deactivated Aryl Chlorides, Chemoselective Arylations, and Room Temperature Reactions. Angewandte Chemie International Edition. 49(24):4071-4074. https://doi.org/10.1002/anie.201000526
2.
Lundgren RJ, Stradiotto M. 2010. Transition-Metal-Catalyzed Trifluoromethylation of Aryl Halides. Angew. Chem. Int. Ed.. 49(49):9322-9324. https://doi.org/10.1002/anie.201004051
3.
Hesp KD, Lundgren RJ, Stradiotto M. 2011. Palladium-Catalyzed Mono-?-arylation of Acetone with Aryl Halides and Tosylates. J. Am. Chem. Soc.. 133(14):5194-5197. https://doi.org/10.1021/ja200009c
4.
Lundgren R, Sappong-Kumankumah A, Stradiotto M. 2010. A Highly Versatile Catalyst System for the Cross-Coupling of Aryl Chlorides and Amines. Chem. Eur. J.. 16(6):1983-1991. https://doi.org/10.1002/chem.200902316
5.
Hesp KD, Stradiotto M. 2010. Stereo- and Regioselective Gold-Catalyzed Hydroamination of Internal Alkynes with Dialkylamines. J. Am. Chem. Soc.. 132(51):18026-18029. https://doi.org/10.1021/ja109192w