MilliporeSigma
Imaging Analysis & Live Cell Imaging
A microscopic image showing multiple cells with distinct blue nuclei surrounded by a network of pink structures, set against a dark background, highlighting the intricate details of cellular components.
Live-cell imaging and analysis allows scientists to study dynamic cellular events in real time to gain unique insights into biology. These techniques are in contrast to PCR, flow cytometry, and immunocytochemistry and tissue staining with antibodies, which analyze cellular events as a snapshot. Today, improvements in cameras, live-cell fluorescent dyes, fluorescent proteins, video compression, and time-lapse microscopy have advanced our ability to visualize and analyze living cells, their intercellular interactions, and subcellular processes with remarkable detail and fidelity.   
Something went wrong, please try again.
Featured Categories
Duolink® Proximity Ligation Assay (PLA) technology. On the left, a diagram depicts protein structures with red and blue interactions. On the right, a fluorescent microscopy image shows a cellular landscape with blue-stained nuclei and green cytoplasmic structures, highlighting protein-protein interactions indicated by a red arrow.
Duolink® Proximity Ligation Assay
Duolink® Proximity Ligation Assay (PLA) technology for detecting and quantifying...
Shop Products
Microscopy image of the Cell Comb scratch assay for cell migration
Cell Migration & Invasion Assays
Explore our assays for mimicking cell behaviors in vivo. Migration, invasion, Boyden chamber, and...
Shop Products
A close-up of a hand in a latex glove holding a transparent plastic device with colored indicators, positioned in front of a computer monitor displaying a vibrant, magnified image of a cell structure with blue and pink hues, set against a turquoise background.
CellASIC® ONIX2 Microfluidics
The CellASIC® ONIX2 Microfluidic System is a powerful, automated platform that allows precise manipulation of the cell culture environment for live-cell imaging and analysis using mammalian, yeast, and bacterial cells.
Shop Products
This image shows two scientists in the MLab with bioreactors in the background.
Cell Culture Media & Buffer
Standard, complex, and specialty cell culture media for a variety of cell lines and applications. We also offer starting buffers and supplements used in custom media development.
Shop Products
Live-cell Imaging Systems
A variety of techniques are used to visualize the complex structures and behavior of cells. In static analyses, cells are fixed and stained, which requires killing the cells and therefore capturing a single instant in cellular time. While static techniques are suitable for some applications, they do not provide insight into cellular activities. Dynamic, live-cell approaches facilitate the visualization of structure, behavior or composition in live cells, without traditional staining. These techniques are ideal for applications that must mimic in vivo conditions, as well as for developing predictive assays for drug screening.
Live-cell imaging systems use advanced optics, environmental control, and reagents that permit long-term studies of mammalian, bacterial, and yeast cells. Culture and imaging systems that also employ microfluidics allow precise control of the culture microenvironment, like flow, gas mixture, and temperature, while limiting physical stress to cells. Live-cell imaging and analysis is suited to a range of applications, including hypoxia, cell migration, 3D cell culture, cytoskeletal dynamics and protein trafficking. Advanced systems automate culture conditions, administration and withdrawal of treatments, and imaging intervals to create images that may be presented as still image or video data.
Live-cell Imaging Reagents
Live-cell imaging media and supplements are designed to protect cells from light-induced cellular damage during time-lapse experiments. These reagents are also formulated to have low autofluorescence and photobleaching, which dramatically enhances the quality of fluorescent live-cell images.
A variety of live-cell staining dyes are available to study the dynamic processes of live cells. Live-cell fluorescent organelle dyes permit the selective staining of specific organelles, such as the cell membrane, nucleus, cytoplasm, mitochondria, lysosomes, endoplasmic reticulum (ER), Golgi apparatus, and cytoskeleton proteins, all without increased cytotoxicity. Using organelle dyes as counterstains in live-cell imaging is also useful in functional studies.
Biosensors with GFP and RFP can be used to detect a specific protein as well as the subcellular location of that protein within live cells by either fluorescent microscopy or time-lapse video capture. Lentiviral vector systems are a popular tool for introducing genes and gene products into cells, with many advantages over chemical-based transfection. Beyond organelle detection, cell-permeable live-cell dyes are used for applications such as apoptosis detection, cell viability, cell health, hypoxia, reactive oxygen species studies, calcium indicator imaging, and neural or stem cell experiments.
An illustration featuring a computer monitor displaying a document with blue text, accompanied by laboratory glassware on the left and a small green plant on the right, all set against a turquoise background.
Looking for More Specific Information?
Visit our document library for data sheets, certificates and technical documentation.
Find Documents