MilliporeSigma
HomeSmall Molecule HPLCMilli-Q® IQ 7000 Ultrapure Water System
Milli-Q® IQ 7000 Ultrapure Water System
An Ultrapure Water Solution Designed with You in Mind
It's intelligent. It's intuitive. It's designed to make your work as pleasant and comfortable as possible — and to maximize lab productivity. Its breakthrough ergonomic design and powerful purification media ensure consistent production of superior-quality ultrapure water. See how the Milli-Q® IQ 7000 water purification system will change the way you work in the lab.
Video: Milli-Q® IQ 7000
See how the Milli-Q® IQ 7000 ultrapure water system can change your daily work in the lab with its ergonomic design, precise dispensing options, high-definition digital touch screen, integrated data management system, digital services, effortless maintenance, and much more.
Simple & Intuitive Dispensing
Dispensing ultrapure water from the Q-POD® has become much easier, precise and effortless with the new “wheel” dispense. You can access a large number of dispensing functions that are useful for day-to-day work in the laboratory.
Large, Interactive Touch Screen
Like a smartphone, the Q-POD® dispenser's interactive touch screen allows you to go anywhere in just a few clicks.
Optimized Lab Space
Protect your working space from clutter! The system is designed for a neat installation and clean lab benches, free of loose tubing and cables. Just interact with your Q-POD® dispenser — the rest of the compact system can easily be integrated in the lab environment. For labs needing more than one ultrapure water point-of-use, up to four Q-POD® dispensers can be connected.
Superior Ultrapure Water — Consistently — to the Trace Level
The Milli-Q® IQ 7000 system provides superior ultrapure water consistently to the trace level with the combination of Jetpore® and IQnano™ resin. Plus, tailored media optimizes performance as purification cartridges can be adapted to your feed water situation, whether extra hard tap water, or Elix® or non-Elix® pure water feed. You can fully rely on the system's water purity for your critical experiments. And with the A10® TOC on-board monitoring, you're spared from uncertainty or misinterpreting results — and also from having to repeat time-consuming and costly analyses.
A Greener Alternative Product
As part of our Design for Sustainability approach to product development, the Milli-Q® IQ 7000 system and its purification kits were developed to deliver high-quality ultrapure water with a reduced environmental footprint versus the previous generation of systems and cartridges.
As a result of these efforts, this system and its kits are identified as Greener Alternative Products thanks to improvements in a range of sustainability characteristics. Indeed, we certify that the Milli-Q® IQ 7000 system uses less electricity, plastic and packaging, and is completely mercury free.
Review the Design for Sustainability (DfS) scorecard to see how the Milli-Q® IQ 7000 system can support your lab's sustainability goals.
Following our Design for Sustainability process, Milli-Q® IQ 7003 and 7010 water systems use less plastic and electricity, and are mercury free.
Advanced Traceability
Advanced data tracking is an integral part of Milli-Q® IQ 7000 system. Dispensing Events ensures a history of all dispense reports, and Daily Quality Measures provides water quality traceability over time. Complete History is a global archive of all system data when more in-depth analysis is required. Traceability is also ensured through the e-Sure tags on system POD-Paks and IPAK cartridges.
Quality & Compliance
Validated and qualified design, combined with our strict manufacturing Quality Assurance system, is your best warranty. We offer a complete range of services, including validation support, which is provided by certified field service engineers using calibrated equipment and a Milli-Q®-specific Qualification Workbook. The Milli-Q® IQ 7000 system is intended to produce ultrapure water that meets or exceeds requirements as described by the ASTM, ISO 3696, CLSI, European Pharmacopoeia and US Pharmacopeia.
Digital Services Simplify System Care and Assure Productivity
MyMilli-Q™ Remote Care is a unique online monitoring and service feature that provides:
  • Remote access to your system and its data 
  • Automatic notifications, by email or SMS, in case of an alert or alarm
  • Faster service with remote diagnostics and repair possible
  • Simplified data management and advanced traceability that eases audit and accreditation processes
MyMilli-Q™ Digital Services Sticker Icon
View the Convenience of Connectivity video on how a connected water purification system can support your uninterrupted lab productivity.
Milli-Q® IQ 7000 for Water Purification
Figure 1.The Milli-Q® IQ 7000 Water Puriffication System is a Greener Alternative Product.
Water Specifications / International Conformity
Feed water requirements
Feed water qualityPretreated water using EDI, DI, RO or distillation technologies
Conductivity at 25 °C< 100 µS/cm (as NaCl)
Total Organic Carbon (TOC)< 50 ppb
Pressure range*Max 6 bar
Temperature5 – 35 °C / 41 – 95 °F
* For pressures between 0 and 0.1 bar, the system will operate, but product flow rate may be lower.
Ultrapure, Type 1 water specifications1 (from Q-POD® dispenser)
Resistivity at 25 °C218. 2 MΩ·cm
TOC3≤ 2 ppb (μg/L)(3), typically ≤ 5 ppb (μg/L)
Particles4No particles with size > 0.22 µm
Bacteria<0.01 cfu/mL (<10 cfu/L)(4,5)
<0.005 cfu/mL (<5 cfu/L)(6)
Pyrogens (endotoxins)5< 0.001 EU/mL
RNases5< 1 pg/mL
DNases5< 5 pg/mL
Proteases5< 0.15 µg/mL
Flow rate0.05 – 2 L/min
1 These values are typical and may vary depending on the nature and concentration of contaminants in the feed water.
2 Resistivity can also be displayed non-temperature-compensated as required by USP.
3 In the appropriate operating conditions, otherwise typically ≤ 5ppb.
4 With Millipak® and Millipak® Gold
5 With Biopak®
6 With Millipak® Gold installed and used in a laminar flow hood
International Regulation Requirements
Declaration of CE and cUL Conformity:
The Milli-Q® IQ 7000 system has been tested by an independent and accredited company for compliance with CE directives related to safety and electromagnetic compatibility. The report can be consulted on demand. In addition, the Milli-Q® IQ 7000 system is built using components and practices recommended by UL and has been cUL marked. The registration can be verified on the UL web site: www.ul.com.