MilliporeSigma
HomeSmall Molecule HPLCAnalysis of Arbutin and Hydroquinone in whitening serum
Analysis of Arbutin and Hydroquinone in whitening serum using a Chromolith® HighResolution RP-18e 2 mm I.D. HPLC column
Anita Piper, R&D Chemist, Lara Celia Bergner
Introduction
Bleaching or skin whitening serums are widely used to reduce melanin content in the skin, and for such products, analytical quality control is required. Hydrochinone is a decomposition product of arbutin which can cause severe contact dermatitis in humans.
This report focuses on the testing of arbutin and hydroquinone in formulated serum products. A Chromolith® HighResolution RP-18 endcapped column 100x2mm was used on an HPLC-UV instrument, and can improve existing methods (e.g. Int. J. Appl. Sci. Eng., 2011.9.4)1.
Two images- on the left bond line chemical structure of arbutin and on the right bond line chemical structure of hydroquinone
EXPERIMENTAL CONDITIONS
Column:
Chromolith® HighResolution RP-18e, 100x2mm (1.52322)
Detection:
Dionex Ultimate 3000 VWD-3400, UV @ 220 nm, micro flow cell (1.4 µL/7 mm)
Mobile phase:
[A] Water, [B] methanol; isocratoc at A/B 90/10 (v/v)
Flow Rate:
91 µL/min
Pressure Drop:
22 bar (319 psi)
Column temp.:
25 °C
Injection volume:
0.5 µL
Samples
Diluent
Water/Methanol 30/70 (v/v)
Standard solution (10 mg/mL)
Accurately weigh 5 mg of each standard into a 50 mL volumetric flask and add 40 mL diluent. Sonicate for 5 min and fill up to mark with diluent. Pipette 5 mL of this solution into a 50 ml volumetric flask and fill up to mark with diluent. Pass through a 0.45 µm syringe filter before injection.
Sample solution
Place 1 g of homogenized serum in a 50 mL volumetric flask, add 30 mL of diluent and sonicate for 5 minutes. Fill up to volume with diluent. Transfer 3 mL of this suspension into a 25 mL volumetric flask and fill up to volume with diluent. The resulting cream sample concentration in the final dilution is 2.4 mg/mL. Pass through a 0.45 µm syringe filter before injection.
Results - Calibration and Repeatability
A Chromatogram obtained for the blank run in the HPLC-UV analysis of arbutin and hydroquinone with intensity on the y-axis and retention time measured in minutes on the x-axis showing a straight line running parallel to the x-axis
Figure 1.Chromatogram blank.
A Chromatogram obtained for a standard solution of arbutin and hydroquinone with 10 µg/mL each in an HPLC-UV analysis with intensity on the y-axis and retention time measured in minutes on the x-axis showing three distinct peaks labeled 1,2, and 3.
Figure 2.Chromatogram standard solution arbutin and hydroquinone each at 10 µg/mL.
Chromatographic Data - Standard Solution (10 µg/mL )
No.
Compound
Retention Time (min)
S/N
Area (mAU*min)
Tailing Factor
1
t0 void volume
2.9
2
Arbutin
4.1
94.3
0.8407
1.4
3
Hydroquinone
5.1
368.4
1.6358
1.8
Specificity Test: The standard solution of arbutin and hydroquinone (each at 10 µg/mL) was injected and the retention time and content of desired analyte determined (Table 1). Repeatability was determined by 5 injections of a serum sample solution (Table 2). Calibration and sensitivity results for arbutin and hydroquinone (calibration range 0.10-15.5 µg/mL for arbutin and 0.10-15.3 µg/mL for hydroquinone with 9 calibrators) are shown in Table 3.
Compound
Retention Time (min)
Area (mAU*min)
Tailing Factor
S/N
1
Arbutin
4.1
0.8407
1.4
141.7
2
Hydroquinone
5.1
1.6358
1.8
488.9
Table 1. Specificity test results (standard solution of arbutin and hydroquinone at 10 µg/mL each)
Sample
Area (mAU*min)
STD 1
5.2146
STD 2
5.2615
STD 3
5.2657
STD 4
5.2224
STD 5
5.3311
Mean
5.2702
Standard Deviation
0.0451
(%) RSD
0.9
Table 2.Repeatability of sample solution (serum with arbutin)
Compound
LOD (µg/mL)
LOQ (µg/mL)
R2
Arbutin
2.7
8.2
0.9992
Hydoquinone
0.5
1.4
0.9994
Table 3.Calibration and sensitivity results for arbutin and hydroquinone (calibration range 0.10-15.5 µg/mL for arbutin and 0.10-15.3 µg/mL for hydroquinone with 9 calibrators).
Calibration curve with mean area on y-axis and concentration in µg/mL on the x-axis, obtained for arbutin measured at different concentrations in the range of 0.10-15.5 µg/mL, showing a straight line starting from 0, following the equation for straight line y=mx+c with m value of 0.0791 and c value of 0.0141, along with R2 value of 0.9992
Figure 3.Calibration curve arbutin.
Calibration curve with mean area on y-axis and concentration in µg/mL on the x-axis, obtained for hydroquinone measured at different concentrations in the range of 0.10-15.3 µg/mL, showing a straight line starting from 0, following the equation for straight line y=mx+c with m value of 0.1490 and c value of 0.0132, along with R2 value of 0.9994
Figure 4.Calibration curve hydroquinone.
The LOD & LOQ in the injected standard solution represent limits for the serum samples of 1.13 mg/g (LOD) and 3.42 mg/g (LOQ) for arbutin 0.21 mg/g (LOD) and 0.58 mg/g (LOQ) for hydroquinone.
Serum Sample Results
A Chromatogram obtained for a serum sample not containing arbutin in an HPLC-UV analysis with intensity on the y-axis and retention time measured in minutes on the x-axis showing only time of the unretained peak (t0) nad no peak for arbutin
Figure 5.Chromatogram serum without arbutin.
A Chromatogram obtained for a serum sample containing arbutin in an HPLC-UV analysis with intensity on the y-axis and retention time measured in minutes on the x-axis showing two peaks labeled, 1 and 2. Peak 1 corresponds to time of the unretained peak (t0) and peak 2 for arbutin
Figure 6.Chromatogram serum containing arbutin
Chromatographic Data - Serum with Arbutin
No.
Compound
Retention Time (min)
S/N
Area (mAU*min)
Tailing Factor
1
t0 void volume
2.9
2
Arbutin
4.1
563.5
4.9694
1.7
3
Hydroquinone
 n.d.
Calculation
ωSample  =         cst/cPr
ωSample  =   (63.00 μg/mL) / (2.4 mg/mL)
ωSample  =    26.25 µg/mg  or 2.63% Arbutin in Serum
where,
ωSample  = Concentration of Arbutin in the sample 
cst    = Concentration calculated from calibration curve (µg/mL)
cPr     = Concentration of serum sample in final dilution (mg/mL)
Conclusion
It was shown, that a Chromolith® HighResolution RP-18e 100x2mm column coupled to UV detection can be utilized for the determination of arbutin and hydrochinone in serum. For arbutin, the resulting limit of detection (LOD) was 2.7 µg/mL, and the limit of quantitation (LOQ) was 8.2 µg/mL in the final dilution sample, representing limits for the serum samples of 1.13 mg/g (LOD) and 3.42 mg/g (LOQ); for hydroquinone, the results were 0.5 µg/mL (LOD) and 1.4 µg/mL (LOQ) in the final dilution sample, representing limits for the serum samples of 0.21 mg/g (LOD) and 0.58 mg/g (LOQ). Due to the excellent permeability and low backpressure of this column type, an analytical HPLC-System with a micro cell could be used.
See more chromatograms for cosmetic applications
Related Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Wang A, Cheng S, Kwan C. 2011. Simultaneous determination of five whitening agents by ion-pair reversed-phase high performance liquid chromatography. [Internet]. Int. J. Appl. Sci. Eng. Available from: https://gigvvy.com/journals/ijase/articles/ijase-201112-9-4-287