Middle-Up Antibody Analysis by RP Chromatography
Benjamin Peters, Gisela Jung
HPLC R&D, Merck KGaA, Darmstadt, Germany
Comparison of Chromolith® WP 300 RP-18, 2 mm I.D., with Other Columns
Section Overview
INTRODUCTION
Analysis of monoclonal antibodies (mAbs) is a difficult analytical challenge that requires a multi-pronged approach. Different modes of chromatography, in addition to different sample preparation strategies, need to be employed for complete characterization. In reversed-phase mode, three levels of analysis are typically performed: top-down (intact), bottom-up (fully digested peptide fragments), and middle-up (larger protein sub-units).
In middle-up analysis, a digestion procedure, such as using dithiothreitol (DTT) or IdeS (a protease) is employed to break the protein into larger fragments. DTT breaks a mAb down into a light chain (LC) fragment and a heavy chain (HC) fragment; IdeS breaks the mAb down into a fragment crystallizable (Fc) fragment and an antigen-binding fragment (Fab). These two approaches lead to a simpler mixture than a full tryptic digest which enables easier identification of variants.
The purpose of this application is to subject the SigmaMAb antibody reference standard and the NISTmAb reference standard to DTT and IdeS digestion and analyze each of the resulting digests by RPC. In addition, a series of columns will be employed to perform the analysis, and the differences in selectivity, efficiency, and peak shape will be examined.
![Antibody Digestion Schemes Digestion scheme of antibody using dithiothreitol (DTT) or IdeS (a protease)](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/antibody-digestion-schemes/antibody-digestion-schemes.jpg)
Figure 1.Digested products of an IgG antibody after being subjected to a) DTT and b) IdeS
Columns: | As Indicated; 10 cm x 2.1 mm I.D., 2.0 µm (SPP), 1.7 µm (FPP), 10cm x 2 mm I.D (Chromolith®) | ||
Detection: | UV @ 220 nm (analytical flow cell; 10 µL) | ||
Mobile phase: | [A] Water (0.1% TFA v/v); [B]Acetonitrile (0.08% TFA v/v) | ||
Gradient: | Time (min) 0 min 1 min 10 min | %B 20 20 45 | |
Sample: | Digested mAb, varied concentration, water (0.1% TFA v/v) | ||
Sample Prep (DTT Method): | 60 µL of 40 mM DTT solution was added to 40 µL mAb and allowed to incubate in a vial at 37 °C for 30 minutes. | ||
Sample Prep (IdeS Method): | 4 µL IdeS and 56 µL water were mixed with 40 µL mAb and allowed to incubate at 37 °C for 30 minutes | ||
Injection Volume: | 1.0 µL | ||
Flow Rate: | 0.38 mL/min | ||
Temperature: | Column: 80 °C Autosampler: 10 °C | ||
Pressure Drop: | As Indicated |
![SigmaMAb DTT Reduction Comparison A chromatogram displaying the SigmaMAb DTT reduction results, with absorbance measured at 220 nm over retention time (0 to 12 minutes). The graph features three different lines representing SPP (red), FPP (blue), and Chromolith® WP 300 RP-18 (yellow), each indicating various pressures. Key peaks are labeled as LC (light chain) and HC (heavy chain).](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/sigmamab-dtt-reduction/sigmamab-dtt-reduction.jpg)
Figure 2.Comparison of separation performance between Chromolith® WP 300, a SPP, and an FPP-packed column analyzing DTT reduced SigmaMAb. Chromolith® performs comparably to these UHPLC columns but with reduced backpressure.
![NIST mAb DTT Reduction Comparison A chromatogram illustrating the NISTmAb DTT reduction results, with absorbance measured at 220 nm over a retention time of 0 to 12 minutes. The graph features three distinct lines representing SPP (red), FPP (blue), and Chromolith® WP 300 RP-18 (yellow), each corresponding to different pressures. Key peaks are labeled as LC (light chain) and HC (heavy chain).](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/nist-mab-dtt-reduction/nist-mab-dtt-reduction.jpg)
Figure 3.Comparison of separation performance between Chromolith®, a SPP, and an FPP-packed column in analyzing DTT reduced NISTmAb. Chromolith® performs comparably to the UHPLC columns but at a significantly lower backpressure.
![IdeS Reduced Sigma MAb Comparison A chromatogram showing the SigmaMAb IdeS reduction results, with absorbance measured at 220 nm across a retention time of 0 to 12 minutes. The graph includes three lines representing SPP (red), FPP (blue), and Chromolith® WP 300 RP-18 (yellow), each associated with different pressures. Key peaks are labeled as FC (fragment crystallizable) and Fab (fragment antigen-binding).](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/sigma-mab-ides-reduction/sigma-mab-ides-reduction.jpg)
Figure 4.Comparison of separation performance between Chromolith®, a SPP, and an FPP-packed column in analyzing IdeS reduced SigmaMAb. Chromolith® is able to separate the Fab and Fc species with high resolution without generating artifacts due to higher pressure, as illustrated by extra peaks and peak broadening in the FPP and SPP, respectively.
![IdeS Reduced NIST mAb Comparison A chromatogram depicting the NISTmAb IdeS reduction results, with absorbance measured at 220 nm over a retention time of 0 to 12 minutes. The graph features three lines representing SPP (red), FPP (blue), and Chromolith® WP 300 RP-18 (yellow), each linked to specific pressures. Key peaks are labeled as LC (light chain) and HC (heavy chain).](/deepweb/assets/sigmaaldrich/marketing/global/images/technical-documents/articles/analytical-chemistry/large-molecule-hplc/nist-mab-ides-reduction/nist-mab-ides-reduction.jpg)
Figure 5.Comparison of separation performance between Chromolith®, a SPP, and an FPP-packed column in analyzing IdeS digested NISTmAb. The Chromolith® column performs just as well as the UHPLC columns.
CONCLUSION
The purpose of this study was to compare the performance of a series of columns in resolving medium-sized fragments of antibodies, after digestion, by RPC. Two different digestion protocols were employed in this study, DTT and IdeS. All three columns achieved excellent resolution between the two protein subunits. For the DTT digest, the Chromolith® WP 300, 2 mm I.D. column achieved equivalent UHPLC performance to the two, dedicated UHPLC columns, indicating that it has equivalent separating performance. As an added benefit, this performance was achieved at ~16% the pressure drop of the FPP column.
For the IdeS digestion, the Chromolith® WP 300, 2 mm I.D. column exhibited better separation performance than the SPP column, as is illustrated by the improved resolution of variants around the Fab peak in both SigmaMAb and NISTmAb. In what appears to be peak splitting occurring on the Fab peak of SigmaMAb on the FPP is actually a new variant generated from the high backpressure seen on the FPP. This artifact is mitigated, without a compromise in efficiency, on the Chromolith® WP 300, 2 mm I.D. column. Further research will examine the composition of this variant.