MilliporeSigma
HomeLarge Molecule HPLCDiscovery C18 and C8 HPLC Columns
Discovery C18 and C8 HPLC Columns
Classic reversed-phases for all general-purpose applications
  • Excellent peak shape and efficiency compared to competitive columns
  • Classic reversed-phase retention and selectivity
  • Excellent, no-bleed character for LC-MS applications
  • C18 is generally more retentive than the C8
Properties C18 C8
Bonded Phase: Octadecylsilane, endcapped Octylsilane, endcapped
Silica: Spherical, high purity (Fe <20; NA <7; Ca <7;
Ti <1; AI <1; Mg <1ppm)
Spherical, high purity (Fe <20; NA <7; Ca <7;
Ti <1; AI <1; Mg <1ppm)
Particle Size: 5 µM 5 µM
Pore Size: 180(Å) 180(Å)
Surface Area: 200m2/g 200m2/g
%C: ~12% ~7.5%
Coverage: ~3µmoles/m2 ~3.4µmoles/m2
C18 structure C8 structure
Discovery C18 and C8 HPLC Columns
Figure 1.Discovery C18 and C8 HPLC Columns
Ordering Information
5µm Discovery HPLC Columns
Sorry, an unexpected error has occurred
Network error: Failed to fetch
2cm Supelguard Cartridges with Discovery Packings
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Discovery C18 and C8 HPLC Columns