MilliporeSigma
HomeqPCRKiCqStart™ One-Step Probe RT-qPCR
KiCqStart™ One-Step Probe RT-qPCR
Product Description
KiCqStart™ One-Step Probe RT-qPCR ReadyMix™ is a ready-to-use, highly sensitive master mix for reverse transcription quantitative PCR (RT-qPCR) of RNA templates using hybridization probe detection chemistries such as TaqMan® 5’-hydrolysis probes on real-time PCR systems (refer to instrument compatibility section to select the appropriate reagent for your machine). First-strand cDNA synthesis and PCR amplification are carried out in the same tube without opening between procedures. It is ideal for highly sensitive quantification of RNA viruses or low abundance RNA targets as well as high throughput gene-expression studies. The system has been optimized to deliver maximum RT-PCR efficiency, sensitivity, and specificity in reduced reaction volumes and fast cycle times.
KiCqStart™ One-Step Probe RT-qPCR ReadyMix™ contains all required components for RT-qPCR except RNA template and probe. It is compatible with all dual-labeled probe chemistries. The reverse transcriptase is an engineered M-MLV with reduced RNase H activity and improved activity and stability at higher temperatures. The use of higher temperatures (50 to 55°C) for the first-strand step of one-step RT-qPCR provides higher specificity for primer annealing and disruption of RNA secondary structure that can interfere with cDNA synthesis.
KiCqStart™ One-Step Probe RT-qPCR ReadyMix™ is highly resistant to PCR inhibitors. A key component of the ReadyMix is an ultra-pure, highly processive, thermostable DNA polymerase that is combined with high avidity monoclonal antibodies. This provides an extremely stringent automatic hot-start that minimizes the potential for primer-dimer and other non-specific PCR artifacts. The light blue color of the inert dye simplifies reaction assembly in white, or clear, plates and helps to minimize pipetting or mixing errors. The dye does not interfere with qPCR performance or affect the stability of the product.
Reagents
2X reaction buffer containing dATP, dCTP, dGTP, dTTP, magnesium chloride, reverse transcriptase, RNase inhibitor protein, hot-start DNA polymerase, ROX reference dye (if applicable), inert blue qPCR dye, and stabilizers
*Refer to Instrument Compatibility Section to select appropriate reagent
**Reaction number based upon a 20µL final reaction volume
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Storage/Stability
KiCqStart™ One-Step Probe RT-qPCR ReadyMix™ is stable for one year when stored in a constant temperature freezer at -20 ºC protected from light. Repeated freezing and thawing does not affect RT-qPCR performance.
Instrument Compatibility
Different real-time PCR systems employ different strategies for the normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagent to your specific instrument. Please consult the following table, or visit our PCR Selection page to find the optimal kit for your instrument platform.
Supplies
Usage Guidelines
  • Primer and Probe Design
    • The design of highly specific primers and probes is a critical parameter for successful one-step RT-qPCR. Use of our OligoArchitect™ program is encouraged in order to minimize the potential for internal secondary structure and complementation at 3’-ends within each primer, the primer pair, and primer/probe combinations.
    • Regions of strong RNA secondary structure should be avoided as this can interfere with primer hybridization and/or impede procession of the reverse transcriptase.
    • For best results, amplicon size should be between 70 and 150 bp. Optimal results may require titration of primer concentration between 300 and 900 nM. A final concentration of 450 nM each primer and 150 nM probe is effective for most applications.
    • The efficacy and efficiency of any primer/probe set should be validated under fast cycling and/or rapid ramp rate protocols before use in qPCR studies.
  • If frozen, thaw KiCqStart™ One-Step Probe RT-qPCR ReadyMix™ on ice. Thoroughly mix by gently vortexing, and then centrifuge to collect contents to the bottom of the tube. Keep on ice prior to use.
  • To maximize specificity, reactions should be assembled on ice. The hot-start DNA polymerase is inactive prior to high temperature activation; however, the reverse transcriptase is active at lower temperatures.
  • First-strand synthesis can be carried out between 42 °C and 55 °C. Optimal results are generally obtained with a 5 to 10-minute incubation at 48 to 50 °C. Longer incubation times for first-strand synthesis (up to 20 min) may be used.
  • We recommend a minimum of 30s incubation at 95 °C to inactivate the RT and activate the hot-start polymerase prior to PCR cycling.
  • The kit is compatible with either fast or standard qPCR cycling protocols. Annealing and or extension temperatures may need to be optimized for a given primer/probe design or fluorogenic probe chemistry. Use the suggested protocol as a starting point. Multiplexed RT-qPCR may benefit from a slightly longer extension time (45 to 60s).
  • Preparation of a reaction cocktail is recommended to reduce pipetting errors and maximize assay precision. Assemble the reaction cocktail with all required components except RNA template and dispense equal aliquots into each reaction tube. Add RNA to each reaction as the final step. Addition of sample as 2 to 5-μL volumes will improve assay precision.
  • Suggested input quantities of template are: 1 pg to 100 ng total RNA; 10 fg to 10 ng poly A(+) RNA; 10 to 1x108 copies viral RNA.
  • After sealing each reaction, vortex gently to mix contents. Centrifuge briefly to collect components at the bottom of the reaction tube.
Reaction Assembly
Final reaction volume may vary from 10 to 50 μL, scale all components proportionally.
After sealing each reaction, vortex gently to mix contents.  Centrifuge briefly to collect components at the bottom of the reaction tube.
RT-qPCR Cycling Protocol
Incubate complete reaction mix in a real-time PCR detection system as follows:
Quality Control
Kit components are free of contaminating DNase and RNase. KiCqStart™ One-Step Probe RT-qPCR ReadyMix™ is functionally tested in RT-qPCR. Kinetic analysis must demonstrate linear resolution over six orders of dynamic range (r2> 0.995) and a PCR efficiency > 90%.
Materials
Loading
Limited Label Licenses
The use of certain types of fluorogenic probes with 5' nuclease assays may be covered by one or more of the following US patents and corresponding patent claims outside the US:  U.S. Patent No. 5,538,848, owned by Life Technologies, Corporation. Purchase of this product does not convey rights to practice the methods claimed in U.S. Patent No. 5,538,848, and a license to practice those methods must be obtained from Life Technologies, 850 Lincoln Center Drive, Forest City, California 94404, or by purchase of fluorogenic probes from an authorized source.  No right under any other patent claim and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser's activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Use of other hybridization probes may be covered by other patents and specific licenses may be required for each technology which is not conveyed by purchase of this product. Consult your probe provider for patent and license information related to each probe chemistry and their use.
Applicable only to products containing Passive Reference The purchase of this product includes a limited, non-transferable right to use the purchased amount of the product to perform Applied Biosystems’ patented Passive Reference Method for the purchaser’s own internal research. No right under any other patent claim and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser’s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. For information about these rights or on obtaining additional rights, please contact [email protected] or Out Licensing, Life Technologies, 5791 Van Allen Way, Carlsbad, California 92008.