MilliporeSigma
HomeWet Chemical AnalysisDispensette® Testing Instructions (SOP)
Dispensette® Testing Instructions (SOP)
bottle-top-dispenser
1. Introduction
The standard ISO DIS 8655 describes both the design and the testing of the bottle-top dispenser. The following Testing Instructions describe how to apply the ISO standard in practice.
We recommend a testing every 3-12 months. This interval may be adjusted to individual requirements. For example, when working very frequently or when using aggressive media, the instrument should be tested more frequently.
These Instructions may also be used as a basis for the supervision of testing devices to DIN EN ISO 9001, DIN EN ISO 10 012 and DIN EN ISO/IEC 17 025.
For the regular examinations required by DIN EN ISO 9001, DIN EN ISO 10 012, DIN EN ISO/IEC 17 025 and the GLP Guidelines, BRAND® additionally provides a calibration service. Your instrument will be returned within a few days together with a test report. For more detailed information, please contact your labware supplier.
2. Preparation for testing and visual examination
2.1. Type and Serial Number
  • Determine instrument type and nominal capacity. ⇒ Enter/ check in Test Record (1).
  • Read Serial Number (embossed at valve block). ⇒ Enter number in Test Record (1).
  • Read customer’s identification, if present. ⇒ Enter identification in Test Record (1).
2.2. Minimal Configuration Dispensette®
  • Dispenser ⇒ Use only manufacturer’s original parts.
  • Discharge tube
  • Filling tube
  • Mounting tool for valves
2.3. Cleaning
  • Rinse instrument with cleaning solution. Then rinse the instrument again several times wich was used. ⇒ Select suitable cleaning solution according to the medium with distilled water.
  • Wipe off the exterior of the Dispensette®.
    ⇒ Unscrew piston and lift it out completely following the instructions of the operating manual. Remove desposits out glass cylinder and piston.
2.4. Visual Examination (Damage, Leakages)
  • Housing ⇒ Enter fillings in Test Record (2).
  • Discharge tube
  • Filling tube
  • Controls
  • Leakage
FaultMeasures
Filling tube or discharging tube bent or damagedPossible safety risk - therefore replace part
(see Operating Manual “Accessories“).
Mechanically damaged connectionsPossible safety risk - therefore place parts or return instrument for repair.
Faulty controlsReturn instrument for repair.
Table 1. Possible faults and resulting measures
2.5. Functional Test
  • Screw the Dispensette® on a bottle filled with distilled or deionised water (according to ISO 3696, at least quality 3).
  • Priming (see Operating Manual). ⇒ A few bubbles up to 1 mm in the glass cylinder are permissible.
  • Check the volume adjustment and that the dispensing piston moves freely. ⇒ The dispensing piston must slide smoothly in the dispensing cylinder.
  • Result ⇒ Enter findings into Test Record (3)
If there is a malfunction of the instrument (e.g., piston difficult to move, sticking valves or leakage) please consult the "Troubleshooting" section of the operating manual.
3. Required Equipment for Testing
  • Dispensette®
  • Bottle (at least 500 mL) filled with distiled or deionised water (according to ISO 3696, at least quality 3, room temperature). ⇒ Match temperature of water and room.
  • Recipient (e.g., Erlenmeyer flask, narrow-mouth) filled with some water. ⇒ Bottom of vessel should be covered.
  • Thermometer with a measuring error of maximum: ± 0.2 °C
  • Place the instrument into the testing room for at least 1 hour (unpacked!). ⇒ Allow instrument to adjust to room temperature.
  • Balance
Selected Volumea of
Appartatus under Test: V
Resulution: mgRepeatability and Linearity: mgStandard Uncertainty of Measurement: m
50 µL < V < 1000 µL0.10.20.2
1 mL < V < 10 mL0.10.20.2
10 mL < V < 100 mL122
Table 2. Recommended specifications. a For practical purposes, the nominal volume may be used to choose the balance.
Traceability of Test Results to National Standards
Through the use of calibrated testing devices (balance and thermometer), the requirement of DIN EN ISO 9001, DIN EN ISO 10012 and DIN EN ISO/IEC 17025 to refer the test to the national standard is fulfilled. The calibration of the balance e.g. can be carried out either by DAkkS calibration or official certification of the balance, or by calibrating the balance with appropiate weights that are traced to the national standard. The calibration of the thermometer can also be carried out by DAkkS calibration or official certification, or by a comparison with thermometers that are traced to the national standard (under defined conditions).
4. Gravimetric test
  1. Determine temperature of the water for testing. ⇒ Enter temperature into Test Record (4).
  2. Set instrument to nominal volume.
  3. Dispense some liquid into the separate vessel. Wipe off the drop on the discharge tube against the wall of the recipient.
  4. Place recipient upon the balance. Tare the balance.
  5. Place recipient under the discharge tube.
  6. Pull up the piston as far as it will go. ⇒ Move slowly and steadily.
  7. Push the piston all the way down. ⇒ Move slowly and steadily.
  8. Wipe dispensing tube at the receiving vessel.
  9. Place receiving vessel upon the balance. Write down the value. ⇒ Enter weighing value into the Test Record (5).
  10. Tare the balance again.
  11. Repeat points 2 to 10 another ten times.
  12. Then carry out ten more weighings each by dispensing 50% resp. 10% of the nominal capacity.
5. Evaluation of gravimetric test results
The values obtained by weighing during the gravimetric test are only the mass values of dispensed volume without correction of air buoyancy. In order to obtain the actual volume, an adjustment calculation to account for water density and air buoyancy must be carried out. To facilitate your calculations and evaluations, we recommend the use of the Windows-compatible calibration software EASYCAL™ from BRAND®.
The following calculations must be carried out:
1. Mean weighing values:
(Example for ten weighing values)
Mathematical equation displaying the calculation of the mean value of a data set, with the mean symbolized by x-bar, equal to the sum of ten data points (x1 to x10) divided by 10.
2. Mean volume:
Mean-volume equatiion. Mathematical equation displaying the relationship V equals X multiplied by Z, where V, X, and Z are variables, with a multiplication dot between X and Z
⇒ For factor Z (Table 4)
⇒ Enter value into Test Record (6a).
3. Standard deviation volume:
Standard-deviation-volume equation.  Mathematical formula for standard deviation, with ‘s’ representing the standard deviation equal to the square root of the sum of squared differences between each data point ‘x’ and the mean ‘x-bar’, divided by 9.
⇒ For factor Z, Table 4
⇒ Enter value into Test Record (6b)
4. Accuracy:
Accuracy equation. Mathematical formula for calculating percentage deviation, represented as A[%], which equals the difference between V measured and V nominal, divided by V nominal, all multiplied by 100.
⇒ Enter value into Test Record (6c)
5. Coefficient of variation:
Coeffiecient equation. Mathematical formula representing the coefficient of variation (CV) as a percentage, where CV is equal to the standard deviation (s) multiplied by 100, all over the square root of the mean (V).
⇒ Enter value into Test Record (6d)
Comparison actual/nominal values:
Use the error limits per Table 5 and 6, or define your own error limits. ⇒ Enter values into Test Record (6e, f).
Result
If calculated values A [%] and CV [%] are smaller than or equal to the error limits, the instrument is in good working order.
If the calculated values are larger than the error limits:
  • Verify if the above instructions have been carefully followed step by step.
  • Observe the suggestions under "Troubleshooting" in the Operating Manual.
  • Calibrate the Dispensette® according the instructions in the Operating Manual.
If these measures are not successful, we offer you the possibility to have your instruments calibrated by the BRAND® Calibration Service.
FaultPossible causesMeasures
Volume too large
  • Drop remaining on the titrating tube.
  • Jerky dispensing.
  • During filling, a drop has already been released.
  • ⇒ Before weighing, wipe off any drop into the receiving vessel. Tare the balance.
    ⇒ Dispense slowly and steadily.
    ⇒ Move piston cautiously toward the upper stop.
    Volume too small
  • Instrument not tight.
  • Air bubbles in the instrument.
  • ⇒ Repeat the functional test valves must be tightened or replaced; discharge tube or filling tube must be mounted properly.
    ⇒ Instrument must be primed
    Other causes
  • Jerky titration
  • Temperature adjustment between instrument, room and water temperature not completed
  • ⇒ Use smooth gentle movements to operate the piston upwards and downwards. Approach the upper and lower stops slowly, so that no drops will fall off the dispensing tube.
    ⇒ Carry out temperature adjustment.
    Table 3. Possible volume faults and resulting measures
    Temperature °C
    Factor Z mL/g
    151.0020
    15.51.0020
    161.0021
    16.51.0022
    171.0023
    17.51.0024
    181.0025
    18.51.0026
    191.0027
    19.51.0028
    201.0029
    20.51.0030
    211.0031
    21.51.0032
    221.0033
    22.51.0034
    231.0035
    23.51.0036
    241.0038
    24.51.0039
    251.0040
    25.51.0041
    261.0043
    26.51.0044
    271.0045
    27.51.0047
    281.0048
    28.51.0050
    291.0051
    29.51.0052
    301.0054
    Table 4. Excerpt from DIN EN ISO 8655 Table refers to 1013 hPa the validity range 980 hPa to 1040 hPa
    Error limits for Dispensette®
    Nominal Volume mLAccuracy Value 6e ≤ ± %Coefficient of Variation Value 6f ≤ %
    Type fix  
    10.50.1
    20.50.1
    50.50.1
    100.50.1
    Type Analog and Digital  
    0.5/0.25/0.051.0/2.0/100.2/0.4/2
    2/1/0.20.5/1.0/50.1/0.2/1
    5/2.5/0.50.5/1.0/50.1/0.2/1
    10/5/10.5/1.0/50.1/0.2/1
    25/12.5/2.50.5/1.0/50.1/0.2/1
    50/25/50.5/1.0/50.1/0.2/1
    100/50/100.5/1.0/50.1/0.2/1
    Table 5. The stated error limits refer to new instruments under optimized testing conditions (qualified operators and standardized ambience conditions).
    Nominal Volume mLError Limits for systematic Error
    ± % [A]
    ± µLError Limits for Random Error
    % [CV]
    µL [s]
    0.051.50.750.40.2
    0.11.51.50.30.3
    0.21.020.30.6
    0.51.050.21
    10.660.22
    20.6120.24
    50.6300.210
    100.6600.220
    250.61500.250
    500.63000.2100
    1000.66000.2200
    Table 6. Excerpt from DIN EN ISO 8655, part 5
    6. Test
    10 single measurements per test volume according to DIN EN ISO 8655. For definition of A, CV and s see sample calculation chapter 5. For checking of partial volumes the values for A [%] and CV [%] which are related to the nominal volume have to be converted.
    For calibration, the error limits to be observed by the operator must be individually defined by the user. For this purpose, the following methods can be applied:
    • If required by the application and if the optimized conditions for measuring are present, the error limits in Table 5 can also be expected in the case of used volumetric instruments in good working order.
    • In analogy to the German regulations for official testing, it is also admissible to apply the limits which are typical for practice. These practice limits correspond to double the limits for official testing. In this case, the values found in Table 5 should be doubled.
    • The user may also define his own individual error limits corresponding to his particular application, and apply these tolerances for the calibration of his instrument.
    The above procedures fulfil the requirements of DIN EN ISO 9001, DIN EN ISO 10012 and DIN EN ISO/IEC 17025.
    7. Calibration Service from BRAND®
    BRAND® offers a full service including calibration and adjustment of Brand- and foreign instruments as well as maintenance and repair if necessary - only for BRAND®- instruments. This saves money and adds the benefit of an independent review organisation for the calibration of the instruments. Further information and the order form for repair- and calibration service are found on www.brand.de.
    7.1. Range of Instruments Covered
    1. Piston-operated pipettes (single- and multichannel)
    2. Bottletop dispensers
    3. Piston burettes (bottle-top burettes)
    4. Hand dispensers
    7.2. Testing According to DIN EN ISO 8655
    At BRAND®, a team of qualified staff, working in temperature and humidity controlled rooms using the most modern balances and calibration software, calibrates Liquid Handling instruments, regardless of their make, according to DIN EN ISO 8655.
    Instruments with adjustable volumes such as HandyStep® electronic, Transferpette®, Transferpette® S, Transferpette® electronic, Transferpette®S -8/-12, Transferpette® S -8/-12. Transferpettor or Dispensette®, Digital Burette or Titrette® are tested at nominal volume, and at 50%, 10% or 20% of nominal volume.
    To document the results, a detailed test report is compiled.
    The BRAND® Calibration Service offers:
    1. Calibration of Liquid Handling instruments, regardless of their make
    2. Detailed calibration certificate
    3. Return of instrument within a few working days
    4. Cost-efficient handling
    8. EASYCAL™ Software – advanced calibration technology
    8.1. For Liquid Handling Instruments and Glass or Plastic Volumetric Instruments
    EASYCAL™ simplifies the tedious task of calibrating liquid handling instruments and glass or plastic volumetric instruments to DIN EN ISO 9001, DIN EN ISO 10012, DIN EN ISO/IEC 17025 and GLP standards. The procedures are outlined step-by-step, and all calculations are performed automatically. Reports are generated to document the calibration. All you need is an analytical balance, a PC Windows® 98/2000, NT (SP6), XP, Vista, 7, printer (optional) and EASYCAL™ software.
    • Suitable for instruments from all manufacturers.
    • Specifications of many instruments preloaded.
    • Testing according to ISO 4787, ISO 8655, etc.
    8.2. Data Entry
    • Connect PC and balance (optional), then start the EASYCAL™ software.
    • 100 common balances are preprogrammed for ease of installation.
    8.3. Documentation
    The calibration certificate contains all important test data on one page.
    9. DAkkS-Calibration Service for Volumetric Instruments at BRAND®
    9.1. DAkkS – Deutsche Akkreditierungsstelle GmbH and DKD
    Two logos: on the left, the ilac-MRA circular logo with concentric circles and central text; on the right, the DAkkS logo with an oval shape and the text ‘DAkkS’ inside, accompanied by the text ‘Deutsche Akkreditierungsstelle D-K-15173/01-00’ below.
    The German Calibration Service (DKD) was founded in 1977 as a joint task of state and economy and constitutes the link between the measuring equipment in industrial and research laboratories, testing institutions and authorities and the national standards of the PTB (the German Institute of Physics and Metrology). It effectively supplements the existing verification system which serves above all the purposes of consumer protection. Based on the legal requirements the DKD Accreditation was successively transformed to the DAkkS Accreditation (Deutsche Akkreditierungsstelle GmbH), starting from 2010. BRAND® has been accredited by the DAkkS since Apr. 23, 2013, with the certificate number D-K-18572-01-00.
    Dakks-A
    9.2. DAkkS-Calibration Certificate and Calibration Symbol
    The DAkkS-Calibration Certificate documents officially on a high level the traceability of measuring results to national and international standards and to international SI-units, as required by standards as DIN EN ISO 9001 and DIN EN ISO/IEC 17025 for monitoring of measuring devices.
    DAkkS-Calibration Certificates are issued when calibrations of an accredited laboratory are requested, when high level calibrations are necessary, when national and international standards are demanded and when reference instruments have to be calibrated.
    9.3. DAkkS – A member in the International Accreditation Network
    DAkkS is a member of the International Laboratory Accreditation Cooperation (ILAC), the highest level international institution for laboratory calibration, and is a signatory to the MRA – Mutual Recognition Agreements.
    The accreditation bodies that are signatories to the ILAC mutual recognition agreements (MRAs) recognize their mutual equivalence, and the equivalence of the calibration certificates issued by those same signatories. Likewise, signatories are obliged generally to promote and recommend recognition of the calibration certificates of other signatories (excluding factory calibration certificates).
    The DAkkS is a member of the EA (European Co-operation for Accreditation), which again is a member of the ILAC (International Laboratory Accreditation Cooperation). A multilateral agreement assures obligatory recognition of the DakkS calibration certificate in a variety of countries.
    9.4. DAkkS-Calibration Laboratory at BRAND®
    In 1998 a calibration laboratory for volumetric instruments at BRAND® has been accredited by the German Calibration Service according to DIN EN ISO/ IEC 17 025. Our calibration laboratory is therefore authorized to issue DAkkS-Calibration Certificates (in several languages) for the volumetric instruments listed below. Furthermore we offer adjustment and – for BRAND® Liquid Handling instruments – repair and maintenance.
    For ordering information on DAkkS-Calibration Certificates for volumetric instruments please consult our General Catalog.
    9.5. Volumetric instruments for which BRAND® issues DAkkS Calibration Certifcates
    BRAND® calibrates the following volumetric instruments (new or already in use and regardless of their make):
    • Piston-operated pipettes, from 0.1 µL to 10 mL
    • Multichannel piston-operated pipettes, from 0.1 µL to 300 µL
    • Piston-operated burettes, from 5 µL to 200 mL
    • Dispensers, Dilutors, from 5 µL to 200 mL
    • Volumetric instruments of glass, calibrated to contain (TC, In) from 1 µL to 10000 mL
    • Volumetric instruments of glass, calibrated to deliver (TD, Ex) from 100 µL to 100 mL
    • Volumetric instruments of plastic, calibrated to contain (TC, In) from 1 mL to 2000 mL
    • Volumetric instruments of plastic, calibrated to deliver (TD, Ex) from 1 mL to 100 mL
    • Density bottles of glass, from 1 cm3 to 100 cm3