MilliporeSigma
UHPLC/MS/MS Analysis of 25 Small Molecule Pharmaceutical Compounds on Ascentis® Express RP-Amide
UHPLC/MS/MS Analysis of 25 Small Molecule Pharmaceutical Compounds on Ascentis® Express RP-Amide application for UHPLC-MS
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Ammonium acetate
Molecular Biology, ≥98%
standard
Product No.
Description
Pricing
Imidacloprid
PESTANAL®, analytical standard
Hydroquinidine hydrochloride
98%
Methapyrilene HCl
analytical standard
4-Aminobiphenyl
analytical standard
Nifedipine
United States Pharmacopeia (USP) Reference Standard
Mirtazapine
United States Pharmacopeia (USP) Reference Standard
Clarithromycin
United States Pharmacopeia (USP) Reference Standard
Losartan potassium
United States Pharmacopeia (USP) Reference Standard
Nefazodone hydrochloride
United States Pharmacopeia (USP) Reference Standard
Atrazine
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Benzoylecgonine solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Amiloride hydrochloride
United States Pharmacopeia (USP) Reference Standard
Hydrochlorothiazide
United States Pharmacopeia (USP) Reference Standard
Lincomycin hydrochloride
United States Pharmacopeia (USP) Reference Standard
Loratadine
United States Pharmacopeia (USP) Reference Standard
Amitriptyline hydrochloride solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Haloperidol solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Imipramine hydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Mianserin hydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Buspirone hydrochloride
United States Pharmacopeia (USP) Reference Standard
Mesoridazine besylate
United States Pharmacopeia (USP) Reference Standard
Nizatidine
United States Pharmacopeia (USP) Reference Standard
Nevirapine anhydrous
United States Pharmacopeia (USP) Reference Standard
Imiquimod
United States Pharmacopeia (USP) Reference Standard
CONDITIONS
column
Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase
[A] 0.1% acetic acid, 5 mM ammonium acetate in water; [B] 5 mM ammonium acetate in acetonitrile
gradient
5% B for 1 min; to 43% B in 6 min; held for 1 min; to 80% B in 4 min; held for 1 min; to 5% B in 1 min; held for 3 min
flow rate
0.4 mL/min
detector
MS, ESI(+) MRM
injection
1 μL
Description
Application
This application highlights the advantage of an embedded polar group (EPG) HPLC phase for the separation of various small molecules. The use of an Ascentis® RP-Amide column allowed for the development of a single UPLC-MS/MS method for the analysis of 25 pharmaceuticals that varied significantly in both chemical and physical properties (log P, pKa, etc.). The presence of an embedded amide group allowed for retention of more polar analytes in comparison to a traditional C18 phase.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany