MilliporeSigma
LC/MS Analysis of Anti-infectives on Ascentis® Express Biphenyl
LC/MS Analysis of Anti-infectives on Ascentis® Express Biphenyl application for LC-MS
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express Biphenyl Column
2.7 μm particle size, L × I.D. 10 cm × 3.0 mm
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
Cinoxacin
p-Coumaric acid
≥98.0% (HPLC)
Dapsone
VETRANAL®, analytical standard
CONDITIONS
column
Ascentis Express Biphenyl, 10 cm x 3.0 mm, 2.7 μm particles (64066-U)
gradient
10 mM ammonium formate (70:30 water:acetonitrile) SW pH 4.00
flow rate
0.6 mL/min
pressure
997 psi (68.7 bar)
column temp.
35 °C
detector
MS, ESI+, XIC
injection
5 μL
sample
300ng/mL in 85:15,  water:methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany