MilliporeSigma
HPLC Analysis of Proteins on Discovery® BIO PolyMA-WAX (Analyte Set #1)
HPLC Analysis of Proteins on Discovery® BIO PolyMA-WAX (Analyte Set #1) application for HPLC
Materials
standard
Product No.
Description
Pricing
Trypsin inhibitor from Glycine max (soybean)
Isoelectric focusing marker, pI 4.6
Trypsin inhibitor from Glycine max (soybean)
BioReagent, For use as a marker in SDS-PAGE
Trypsin inhibitor from Glycine max (soybean)
lyophilized powder
Trypsin inhibitor from Glycine max (soybean)
lyophilized powder
Trypsin inhibitor from Glycine max (soybean)
powder, BioReagent, suitable for cell culture
Myoglobin from equine heart
≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
Myoglobin from equine heart
Isoelectric focusing marker, pI (1) 6.8, (2) 7.2
Myoglobin from human heart
≥95% (SDS-PAGE), buffered aqueous glycerol solution
Trypsin inhibitor from bovine pancreas
Type I-P, essentially salt-free, lyophilized powder
Conalbumin from chicken egg white
Iron complex
Conalbumin from chicken egg white
suitable for cell culture, BioReagent
Conalbumin from chicken egg white
Substantially iron-free
Trypsin inhibitor from Phaseolus limensis (lima bean)
Type II-L, crude powder
Trypsin inhibitor from Glycine max (soybean)
solution, sterile-filtered, suitable for, suitable for cell culture
Myoglobin from equine skeletal muscle
95-100%, essentially salt-free, lyophilized powder
Trypsin inhibitor from chicken egg white
Type III-O (free of ovoinhibitor)
Trypsin inhibitor from chicken egg white
Type II-O, Partially purified ovomucoid, containing ovoinhibitor
Trypsin inhibitor from Glycine max (soybean)
lyophilized powder, ~10000 U/mg
Myoglobin from equine skeletal muscle
BioUltra, 95-100% (SDS-PAGE)
Trypsin Inhibitor from Glycine max (soybean)
BioUltra, lyophilized powder, ≥95% (Kunitz inhibitor, SDS-PAGE)
CONDITIONS
column
Discovery BIO PolyMA-WAX, 5 cm x 4.6 mm, 5 μm particles (59602-U)
mobile phase
[A] 10 mM Tris, pH 8.0 with HCl; [B] 10 mM Tris, 0.5 M NaCl, pH 8.0 with HCl; 5 to 100% B in 15 min. (linear)
flow rate
0.5 mL/min
detector
UV, 280 nm
injection
10 μg trypsin inhibitor, 5 μg conalbumin, myoglobin or 10 μg ribonuclease A, 5 μg cytochrome c, a-chymotrypsinogen A
Description
Analysis Note
The large pore diameter of Discovery BIO PolyMA-SCX and PolyMA-WAX particles allows full access to small peptides and very large proteins and protein aggregates. This Figure shows the separation of proteins that vary from 18 to 80 kDA.
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany