MilliporeSigma
LC/MS (TOF) Analysis of Opioids and Opiate-Dependence Management Drugs on Ascentis® Express HILIC
LC/MS (TOF) Analysis of Opioids and Opiate-Dependence Management Drugs on Ascentis® Express HILIC application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
6-Acetylmorphine, 1.0 mg/mL solution
1 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Codeine solution
1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Hydromorphone solution
1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
6-Acetylcodeine solution
1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Hydrocodone solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Morphine solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Oxycodone solution
1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis Express HILIC, 5 cm x 2.1 mm I.D., 2.7 μm particles (53934-U)
mobile phase
2 mM ammonium formate in 90:10 (v:v) acetonitrile:water
flow rate
0.3 mL/min
column temp.
55 °C
detector
TOF/MS
injection
2 μL
sample
400 ng/mL in acetonitrile
Description
Analysis Note
The rapid analysis by HPLC with TOF-MS detection of seven opioids is shown here using an Ascentis Express HILIC column. Cerilliant CRMs were used to ensure reliable MS identification and quantification.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany