MilliporeSigma
LC/MS Analysis of Vaccinium Anthocyanidins on Ascentis® Express C18 at Elevated Temperature and Flow
LC/MS Analysis of Vaccinium Anthocyanidins on Ascentis® Express C18 at Elevated Temperature and Flow application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
standard
Product No.
Description
Pricing
Pelargonidin chloride
Delphinidin chloride
analytical standard
Peonidin 3-O-glucoside chloride
analytical standard
Cyanidin chloride
≥95% (HPLC)
Malvidin chloride
analytical standard
CONDITIONS
column
Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 μm particles (53823-U)
mobile phase
[A] 0.1% formic acid in water; [B] 0.1% formic acid in 75:25 (v/v) acetonitrile:water; (78:22, A:B)
flow rate
0.6 mL/min
pressure
5440 psi (375 bar)
column temp.
35 °C
detector
ESI(+), TOF
injection
1 μL
sample
20 μg/mL in 0.1% formic acid
Description
Analysis Note
Fruits of the genus Vaccinium are popular sources for anthocyanidin antioxidants. This genus includes the commonly known blueberry, huckleberry, and bilberry.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany