MilliporeSigma
LC/MS Analysis of Polypeptide Drugs on Ascentis® Express RP-Amide
LC/MS Analysis of Polypeptide Drugs on Ascentis® Express RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Ammonium acetate
LiChropur, eluent additive for LC-MS
standard
Product No.
Description
Pricing
Oxytocin
United States Pharmacopeia (USP) Reference Standard
Desmopressin
European Pharmacopoeia (EP) Reference Standard
Eptifibatide acetate
≥98% (HPLC)
Bivalirudin trifluoroacetate salt
≥97% (HPLC)
CONDITIONS
column
Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase
[A] 10 mM ammonium acetate in water, pH 4.2; [B] 10 mM ammonium acetate in 90:10 (v/v) acetonitrile:water, pH 4.7
gradient
held at 20% B for 0.1 min; 20 to 80% B in 2.9 min; 80 to 20% B in 0.01 min; held at 20% B for 0.99 min
flow rate
0.3 mL/min
column temp.
35 °C
detector
TOF, ESI(+), m/z 100-2500
injection
3 μL
sample
1 μL/mL in 10mM ammonium formate, 80:20, acetonitrile:water
Description
Analysis Note
This application demonstrates the suitability of the Ascentis Express RP Amide for the analysis of polypeptide drugs.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany