MilliporeSigma
LC-MS Analysis of Explosives on Ascentis® Express C18
LC-MS Analysis of Explosives on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 3 mm
standard
Product No.
Description
Pricing
HMX solution
1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®
RDX solution
1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®
2,4,6-Trinitrotoluene solution
1000 μg/mL in acetonitrile, ampule of 1.2 mL, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis Express C18, 10 cm x 3.0 mm I.D., 2.7 μm particles (53814-U)
mobile phase
[A] methanol; [B] water; (50:50, A:B)
flow rate
0.9 mL/min
column temp.
50 °C
detector
MS: APCI(-)
injection
5 μL
sample
100 ng/mL in 50:50, methanol:water
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany