MilliporeSigma
HPLC (Size Exclusion) Analysis of Proteins on Zenix®-C SEC-300, Effect of DTT and EDTA
HPLC (Size Exclusion) Analysis of Proteins on Zenix®-C SEC-300, Effect of DTT and EDTA application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Zenix®-C SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel
CONDITIONS
column
Zenix-C SEC-300, 30 cm x 7.8 mm I.D., 3 μm particles (Z777092)
mobile phase
(A) 150 mM sodium phosphate buffer, pH 7.0 (B) 150 mM sodium phosphate buffer, pH 7.0 with 2 mM DTT, 0.1 mM EDTA (C) 150 mM sodium phosphate buffer, pH 7.0 with 2 mM DTT, 0.1 mM EDTA
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 280 nm
injection
10 μL
sample
proteins
Description
Legal Information
Zenix is a registered trademark of Sepax Technologies