MilliporeSigma
HPLC Separation of D-sphingosine and related compounds on Ascentis® Express C18
HPLC Separation of D-sphingosine and related compounds on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
standard
Product No.
Description
Pricing
D-erythro-Dihydrosphingosine
≥98%
CONDITIONS
column
Ascentis Express C18, 10 cm X 2.1 mm, 2.7um particles (53823-U)
mobile phase
(A) Water; (B) Acetonitrile, each with 0.1% formic acid/10 mM ammonium formate
gradient
70% B for 8 min, 0.5 min to 90%B, hold for 1.5 min, back to 70% in 0.5 min, hold for 3.5 min
flow rate
0.3 mL/min
pressure
3900 psi (269 bar)
column temp.
35 °C
detector
ESI-MS (+)
detector
D-erythro-Sphingosine 300.3/282.3  MRM(+) ; L-threo-Sphingosine 300.3/282.3 MRM(+) ; D-erythro-Dihydrosphingosine 302.3/284.3 MRM(+)
injection
1 μL
sample
10 ng/mL in 50% methanol/0.1% formic acid
Description
Analysis Note
HPLC Separation of D-erythro- and L-threo-Sphingosine and its related compounds.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany