MilliporeSigma
HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO Wide Pore C18
HPLC Analysis of Cytochrome c Species Variants on Discovery® BIO Wide Pore C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® BIO Wide Pore C18 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
Cytochrome c from pigeon breast muscle
≥95% based on Mol. Wt. 12,173 basis
Cytochrome c from bovine heart
≥95% based on Mol. Wt. 12,327 basis
Cytochrome c from bovine heart
≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Cytochrome c from equine heart
BioReagent, suitable for GFC marker
Cytochrome c from equine heart
≥95% based on Mol. Wt. 12,384 basis
Cytochrome c from equine heart
≥95% (SDS-PAGE)
Cytochrome c from Saccharomyces cerevisiae
≥85% based on Mol. Wt. 12,588 basis
Cytochrome c from human heart
≥95% (SDS-PAGE), lyophilized powder
Cytochrome c from bovine heart
≥95% (GE)
Cytochrome c from equine heart
BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
CONDITIONS
column
Discovery BIO Wide Pore C18, 15 cm × 4.6 mm I.D., 5 μm particles (568222-U)
mobile phase
[A] (0.1% TFA in water):(0.1% TFA in acetonitrile); (70:30), [B] (0.1% TFA in water):(0.1% TFA in acetonitrile), 64:36
gradient
0-100% B in 30 min
flow rate
1 mL/min
column temp.
ambient
detector
UV, 220 nm
injection
12 μL each at 0.8 mg/mL in 0.1%TFA
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany