MilliporeSigma
HPLC Analysis of Water Soluble Vitamins on Ascentis® RP-Amide
HPLC Analysis of Water Soluble Vitamins on Ascentis® RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® RP-Amide HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
Thiamine hydrochloride
reagent grade, ≥99% (HPLC)
Pyridoxine
≥98%
Ascorbic acid
United States Pharmacopeia (USP) Reference Standard
Niacin
United States Pharmacopeia (USP) Reference Standard
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
25 mM potassium phosphate monobasic, pH 3.5
flow rate
1.0 mL/min
column temp.
30 °C
detector
UV, 230 nm
injection
10 μL
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany