HPLC Analysis of Vitamins, Fat Soluble (A and E), on Discovery® C8
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C8, 15 cm × 4.6 mm I.D., 5 μm particles (59353-U)
mobile phase
[A] acetonitrile: [B] water (90:10 v/v, A:B)
flow rate
2 mL/min
detector
UV, 290 nm
injection
10 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany