HPLC Analysis of Tramadol Enantiomers on Astec® CYCLOBOND I 2000 DMP
![HPLC Analysis of Tramadol Enantiomers on Astec® CYCLOBOND I 2000 DMP application for HPLC](/deepweb/assets/sigmaaldrich/product/images/105/918/0fcc5f97-e663-4b3b-ac59-fedae8ffda47/800/0fcc5f97-e663-4b3b-ac59-fedae8ffda47.jpg)
CONDITIONS
column
CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase
[A] 0.1% triethylamine acetate, pH 5.0; [B] acetonitrile; (80:20, A:B)
flow rate
1.2 mL/min
column temp.
23 °C
detector
UV, 254 nm
injection
5 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null