MilliporeSigma
HPLC Analysis of Testosterone and 1-Testosterone on Ascentis® Express C18
HPLC Analysis of Testosterone and 1-Testosterone on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 4.6 mm
CONDITIONS
column
Ascentis Express C18, 10 cm x 4.6 mm I.D., 2.7 μm particles (53827-U)
mobile phase
[A] water; [B] methanol; (20:80, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 240 nm
injection
5 μL
sample
50 μg/mL in 10:90 water:methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany