MilliporeSigma
HPLC Analysis of Substituted Poly Aromatic Hydrocarbons using Ascentis® Express Biphenyl
HPLC Analysis of Substituted Poly Aromatic Hydrocarbons using Ascentis® Express Biphenyl application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express Biphenyl Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
standard
Product No.
Description
Pricing
Uracil
≥99.0%
Acenaphthene
99%
CONDITIONS
column
Ascentis Express Biphenyl, 10 cm x 2.1 mm, 2.7 μm particles (64065-U)
mobile phase
[A] Water; [B] Methanol; (30:70 A:B)
flow rate
0.5 mL/min
pressure
6200 psi (427 bar)
column temp.
35 °C
detector
UV, 254 nm
injection
1 μL
sample
Uracil (10 μg/mL), Acenaphthenol (250 μg/mL), Acenaphthenone (20 μg/mL),
  Acenaphthene (500 μg/mL), Prepared in 70:30 A:B
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express Biphenyl for the separation of substituted PAHs. It shows the suitability of this phase for separations of extended aromatics and their oxygenated counter parts. These compounds are potentional metabolites, used as synthetic intermediates, and in the study of dihydrodiol dehydrogenase activity in celluar systems.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany