MilliporeSigma
HPLC Analysis of Strigol Enantiomers on Astec® CYCLOBOND I 2000 RSP
HPLC Analysis of Strigol Enantiomers on Astec® CYCLOBOND I 2000 RSP application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CYCLOBOND I 2000 RSP Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CYCLOBOND I 2000 RSP, 25 cm x 4.6 mm I.D., 5 μm particles (20324AST)
mobile phase
[A] 0.1% triethylamine, pH 4.1; [B] acetonitrile; (75:25, A:B)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 240 nm
injection
5 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null