MilliporeSigma
HPLC Analysis of Ruelene (Cruformate) Enantiomers on Astec® CYCLOBOND I 2000
HPLC Analysis of Ruelene (Cruformate) Enantiomers on Astec® CYCLOBOND I 2000 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CYCLOBOND I 2000 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] acetonitrile; [B] acetic acid; [C] TEA; (100:0.3:0.2, A:B:C)
flow rate
0.6 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
Ruelene (cruformate), 1 mg/mL in acetonitrile
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null