MilliporeSigma
HPLC Analysis of Proteins on BIOshell A160 Peptide C18
HPLC Analysis of Proteins on BIOshell A160 Peptide C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express Peptide ES-C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 2.1 mm
standard
Product No.
Description
Pricing
Trypsin inhibitor from Glycine max (soybean)
lyophilized powder
Adrenomedullin 52 human
≥90% (HPLC), powder
Ribonuclease A from bovine pancreas
for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Lysozyme from chicken egg white
powder (crystalline), 70000-140000 U/mg
Superoxide Dismutase from bovine erythrocytes
lyophilized powder, ≥3,000 units/mg protein, Protein ≥95 % by biuret
Ovalbumin-Atto 488 conjugate
suitable for fluorescence
Cytochrome c equine
recombinant, expressed in E. coli
Carbonic Anhydrase from bovine erythrocytes
≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
CONDITIONS
column
BIOshell A160 Peptide C18, 15 cm x 2.1 mm I.D., 2.7 μm particles (66905-U)
mobile phase
[A] 75:25, 0.1% TFA in water: 0.08% TFA in acetonitrile; [B] 25:75, 0.1% TFA in water: 0.08% TFA in acetonitrile
gradient
held at 0% B for 0.5 min; 0 to 100% B in 7 min
flow rate
0.3 mL/min
pressure
4743 psi (327 bar)
column temp.
35 °C
detector
UV, 220 nm
injection
1 μL
sample
0.2 - 0.6 g/L in mobile phase A
Description
Legal Information
null