MilliporeSigma
HPLC Analysis of Proline Enantiomers on Astec® CHIROBIOTIC® T, Fast Analysis
HPLC Analysis of Proline Enantiomers on Astec® CHIROBIOTIC® T, Fast Analysis application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
water
flow rate
0.65 mL/min
column temp.
25 °C
detector
UV, 210 nm
injection
10 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC