HPLC Analysis of Polar and Non-Polar Analytes on Ascentis® Express C18, Column Test Mixture
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express C18, 5 cm × 3.0 mm I.D. 2.7 μm particles (53811-U)
mobile phase
80% acetonitrile
flow rate
1.2 mL/min
pressure
3700 psi (255 bar)
column temp.
35 ° C
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany