MilliporeSigma
HPLC Analysis of Phenolics from Rosemary on Ascentis® Express C8
HPLC Analysis of Phenolics from Rosemary on Ascentis® Express C8 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C8, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 3 mm
standard
Product No.
Description
Pricing
Carnosic acid
analytical standard
Carnosol
analytical standard
Rosmarinic acid
≥98% (HPLC), from Rosemarinus officinalis L.
CONDITIONS
column
Ascentis Express C8, 5 cm x 3 mm I.D., 2.7 μm particles (53848-U)
mobile phase
[A] 0.1% formic acid in water; [B] 0.1% formic acid in acetonitrile
gradient
15 to 100% B in 8.5 min
flow rate
0.6 mL/min
column temp.
35 °C
detector
UV, 214 nm
injection
2 μL
sample
100 mg/L in 75 μM EDTA, 1.5 mM ascorbate, 15% acetonitrile
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express C8 for the efficient separation of phenolics from rosemary.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany