MilliporeSigma
HPLC Analysis of Paraquat and Diquat on Ascentis® Express HILIC (UV Detection)
HPLC Analysis of Paraquat and Diquat on Ascentis® Express HILIC (UV Detection) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
standard
Product No.
Description
Pricing
Paraquat dichloride hydrate
PESTANAL®, analytical standard
CONDITIONS
column
Ascentis Express HILlC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase
[A] 200 mM ammonium TFA; [B] acetonitrile; (20:80, A:B)
flow rate
0.4 mL/min
column temp.
60 °C
detector
UV, 257 nm (paraquat) and 308 nm (diquat)
injection
1 μL
sample
50 mg/L in mobile phase
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany