MilliporeSigma
HPLC Analysis of Pain Relievers on Ascentis® Express C18
HPLC Analysis of Pain Relievers on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 5 μm HPLC Column
5 μm particle size, L × I.D. 10 cm × 4.6 mm
standard
Product No.
Description
Pricing
Caffeine
meets USP testing specifications, anhydrous
Aspirin
meets USP testing specifications
Acetaminophen
meets USP testing specifications, 98.0-102.0%, powder
CONDITIONS
column
Ascentis Express C18, 10 cm x 4.6 mm I.D., 5 μm particles (50536-U)
mobile phase
[A] water with 3% acetic acid; [B] methanol with 3% acetic acid; (79:21, A:B)
flow rate
2.0 mL/min
pressure
3290 psi (227 bar)
column temp.
45 °C
detector
UV,275 nm
injection
10 μL
sample
100 - 360 μg/mLin mobile phase
Description
Analysis Note
This application shows the improvements in efficiency that may be obtained for USP methods using modern Fused-Core technology.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany