MilliporeSigma
HPLC Analysis of Nucleobases using TSKgel® SuperSW mAb HTP column in HILIC mode
HPLC Analysis of Nucleobases using TSKgel® SuperSW mAb HTP column in HILIC mode application for HPLC
Materials
analytical column
Product No.
Description
Pricing
TSKgel® Amide-80 Guard Cartridge
phase carbamoyl, L × I.D. 1 cm × 2 mm, 5 μm particle size, pkg of 3 ea
TSKgel® SuperSW mAb HTP Guard Column
phase diol, L × I.D. 2 cm × 3 mm, 4 μm particle size
standard
Product No.
Description
Pricing
Adenine
≥99%
Uracil
≥99.0%
Cytosine
≥99%
Cytidine
99%
CONDITIONS
column
TSKgel SuperSW mAb HTP 15 cm x 4.6 mm I.D., 4 μm particles (822855)
column
TSKgel Amide-80 10 cm x 2 mm I.D., 5 μm particles (819695)
mobile phase
HILIC mode: [A]: acetonitrile (HILIC mode); [B]: 15 mmol/L ammonium bicarbonate, pH 7.4 (HILIC mode)
mobile phase
SEC conditions: 100 mmol/L phosphate/100 mmol/L sodium sulfate, pH 6.7 + 0.05% NaN3 (SEC mode)
gradient
Isocratic
flow rate
0.4 mL/min
column temp.
Ambient
detector
UV, 280 nm
injection
1 μL
sample
uracil (1.5 mg/mL), adenine (1.5 mg/mL), cytosine (1.5 mg/mL), cytidine (1.5 mg/mL)
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation