MilliporeSigma
HPLC Analysis of Nonvolatile Pesticides in Water on SUPELCOSIL LC-18-DB after SPE using Supelclean ENVI-Carb
HPLC Analysis of Nonvolatile Pesticides in Water on SUPELCOSIL™ LC-18-DB after SPE using Supelclean™ ENVI-Carb™ application for SPE, application for HPLC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOSIL LC-18-DB HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
SPE tube or plate
Product No.
Description
Pricing
Supelclean ENVI-Carb SPE Tube
bed wt. 250 mg, volume 3 mL, pk of 54
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
sample/matrix
water spiked with pesticides at 10-50 μg/L
SPE tube/cartridge
Supelclean ENVI-Carb, 250 mg/3 mL (57088)
condition
5 mL methylene chloride:methanol (80:20); 1 mL methanol; 10 mL 2% acetic acid in water (keep bed moist until sample addition)
sample addition
100 mL sample at 5 mL/min
drying
1 minute with vacuum
elution
0.8 mL methanol; 2 x 35 mL methylene chloride:methanol (80:20)
eluate post-treatment
dry eluate to 500 μL under gentle nitrogen stream, reconstitute to 1 mL with methanol
column
SUPELCOSIL LC-18-DB, 25 cm × 4.6 mm I.D., 5 μm particles (58355-U)
mobile phase
[A] water:acetonitrile (90:10), [B] acetonitrile; gradient 80% A for 5 min then to 30% A over 30 min
flow rate
1.5 mL/min
detector
UV, 220 nm
injection
20 μL
Description
Legal Information
ENVI-Carb is a trademark of Sigma-Aldrich Co. LLC
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC