MilliporeSigma
HPLC Analysis of Metoprolol Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Metoprolol Enantiomers on Astec® CHIROBIOTIC® T2 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T2 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Application
Product No.
Description
Pricing
HPLC Analysis of Metoprolol Enantiomers on Astec® CHIROBIOTIC® T2 (MS Mobile Phase)
application for HPLC
HPLC Analysis of Metoprolol Enantiomers on Astec® CHIROBIOTIC® V
application for HPLC
standard
Product No.
Description
Pricing
(±)-Metoprolol (+)-tartrate salt
≥98% (titration), powder
CONDITIONS
column
CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 μm particles (16024AST)
mobile phase
0.1 wt% ammonium trifluoroacetic acid in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC