MilliporeSigma
HPLC Analysis of Methorphan Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of Methorphan Enantiomers on Astec® CHIROBIOTIC® V2 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V2 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Application
Product No.
Description
Pricing
HPLC Analysis of Dextromethorphan Enantiomers on Astec® CHIROBIOTIC® V2
application for HPLC
standard
Product No.
Description
Pricing
Dextromethorphan hydrobromide
meets USP testing specifications
Dextromethorphan hydrobromide monohydrate
tested according to Ph. Eur.
CONDITIONS
column
CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 μm particles (15024AST)
mobile phase
0.05 wt% ammonium trifluoroacetic acid in methanol
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
1 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC