MilliporeSigma
HPLC Analysis of mAb on Antibodix® WCX-NP10, Competitive Comparison
HPLC Analysis of mAb on Antibodix® WCX-NP10, Competitive Comparison application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Antibodix® WCX-NP10 Weak Cation Exchange Column
L × I.D. 25 cm × 4.6 mm, 10 μm particle size
CONDITIONS
column
Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 μm particles (Z777272)
mobile phase
(A) 10 mM phosphate buffer, pH 7.5; (B) A + 100 mM NaCl
gradient
15 to 55% B in 30 min
flow rate
0.8 mL/min
column temp.
ambient
detector
UV, 214 nm
injection
5 μL
sample
monoclonal antibody
Description
Legal Information
Antibodix is a registered trademark of Sepax Technologies