MilliporeSigma
HPLC Analysis of Hydrophobic Bases on Ascentis® RP-Amide
HPLC Analysis of Hydrophobic Bases on Ascentis® RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® RP-Amide HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
[A] 25 mM ammonium phosphate, pH 7; [B] methanol; (35:65, A:B)
flow rate
1.2 mL/min
column temp.
35 °C
detector
UV, 230 nm
injection
10 μL
sample
N05158
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany