MilliporeSigma
HPLC Analysis of Glutamine Enantiomers on Astec® CHIROBIOTIC® T (23 °C)
HPLC Analysis of Glutamine Enantiomers on Astec® CHIROBIOTIC® T (23 °C) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
L-Glutamine
meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
L-Glutamine
L-Glutamine
γ-irradiated, BioXtra, suitable for cell culture
D-Glutamine
≥98% (HPLC)
L-Glutamine solution
200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
L-Glutamine solution
200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
L-Glutamine
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] ethanol; (50:50, A:B)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 205 nm
injection
5 μL
sample gas
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC