MilliporeSigma
HPLC Analysis of Furans on Ascentis® Express Phenyl-Hexyl
HPLC Analysis of Furans on Ascentis® Express Phenyl-Hexyl application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express Phenyl-Hexyl, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 3 mm
standard
Product No.
Description
Pricing
5-Hydroxymethyl-2-furaldehyde
99%
Furfuryl alcohol
analytical standard
CONDITIONS
column
Ascentis Express Phenyl-Hexyl, 5 cm x 3.0 mm I.D., 2.7 μm particles (53342-U)
flow rate
0.4 mL/min
column temp.
35 °C
detector
UV, 230 nm
injection
1 μL
sample
20 μg/mL in 80:20, water: acetonitrile
mobile phase
[A] water; [B] acetonitrile; (80:20, A:B)
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express Hexyl-Phenyl for the efficient separation of furans as listed in ASTM method D5837-99.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany