MilliporeSigma
HPLC Analysis of Epinephrine, Epinastine, and Benzalkonium Chlorides on Ascentis® Express HILIC
HPLC Analysis of Epinephrine, Epinastine, and Benzalkonium Chlorides on Ascentis® Express HILIC application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis Express HILIC, 15 cm x 4.6 mm I.D., 2.7 μm particles (53981-U)
mobile phase
[A] 50 mM ammonium acetate; [B] acetonitrile; (10:90, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
ESI(+), XIC m/z 304 & 332 (BAC), 250 (epinastine), 180 (epinephrine)
injection
10 μL
sample
10 mg/L ea. in acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany