MilliporeSigma
HPLC Analysis of CYP3A Inhibitors on Ascentis® Express RP-Amide
HPLC Analysis of CYP3A Inhibitors on Ascentis® Express RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
mobile phase component
Product No.
Description
Pricing
Ammonium formate
eluent additive for LC-MS, LiChropur, ≥99.0%
standard
Product No.
Description
Pricing
Nefazodone hydrochloride
≥98% (HPLC), solid
Sertraline hydrochloride
≥98% (HPLC)
Fluoxetine hydrochloride
United States Pharmacopeia (USP) Reference Standard
Fluvoxamine maleate
United States Pharmacopeia (USP) Reference Standard
Lovastatin
Pharmaceutical Secondary Standard; Certified Reference Material
CONDITIONS
column
Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase
[A] 20 mM ammonium formate in 50:50 (v/v) water:acetonitrile
flow rate
0.6 mL/min
column temp.
35 °C
detector
ESI(+), m/z 100-600
injection
1 μL
Description
Analysis Note
This application demonstrates the suitability of the Ascentis Express RP Amide for the analysis of CYP3A Inhibitors.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany