MilliporeSigma
HPLC Analysis of Carotenes on Ascentis® Express RP-Amide, Methanol Mobile Phase
HPLC Analysis of Carotenes on Ascentis® Express RP-Amide, Methanol Mobile Phase application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
β-Carotene
synthetic, ≥95% (HPLC), crystalline
CONDITIONS
column
Ascentis Express RP-Amide, 15 cm x 4.6 mm I.D., 2.7 μm particles (53931-U)
mobile phase
methanol
flow rate
1 mL/min
pressure
1653 psi (114 bar)
column temp.
30 °C
detector
UV-VIS, 454 nm
injection
5 μL
sample
V8 Splash® fruit juice was extracted with hexane, the extract dissolved 1:1 into ethanol for HPLC injection
Description
Analysis Note
Analysis was conducted using juice extract, detection via UV.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
V8 Splash is a registered trademark of CSC Brands, LP