HPLC Analysis of Carboxymethylated Cytochrome c (Chicken) Tryptic Digest on Discovery® BIO Wide Pore C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery BIO Wide Pore C18, 15 cm × 2.1 mm I.D., 3 μm particles (567202-U)
mobile phase
[A] water/0.1% TFA; [B] (water/0.1%TFA):(acetonitrile/0.1% TFA); 50:50
flow rate
0.20 mL/min
column temp.
35 °C
detector
UV, 215 nm
injection
5 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany