MilliporeSigma
HPLC Analysis of Caffeine and Metabolites on Ascentis® RP-Amide
HPLC Analysis of Caffeine and Metabolites on Ascentis® RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® RP-Amide HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
[A] 0.1% ammonium acetate in water, pH 4.5 with acetic acid; [B] acetonitrile
gradient
2 to 8% B in 3 min; to 20% B in 9 min; held for 2 min; to 2% B in 0.1 min; held for 0.9 min
flow rate
1.0 mL/min
column temp.
30 °C
detector
UV, 275 nm
injection
10 μL
sample
as indicated in 0.1% ammonium acetate in water (pH 4.5 with acetic acid)
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany