MilliporeSigma
HPLC Analysis of Bovine Liver Extract on Mono Q® 5/50 GL
HPLC Analysis of Bovine Liver Extract on Mono Q® 5/50 GL application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Mono Q® 5/50 GL
10 μm particle size, L × I.D. 5 cm × 5 mm
CONDITIONS
column
Mono Q 5/50 GL, 5 cm x 5 mm I.D., 10 μm particles (5-4807)
mobile phase
[A] 50 mM diethanolamine, pH 8.8; [B] 50 mM diethanolamine + 1.0M sodium chloride, pH 8.8
flow rate
1.0 mL/min
detector
UV, 280 nm
injection
100 μL 1.6 mg protein
Description
Legal Information
Mono Q is a registered trademark of Cytiva