MilliporeSigma
HPLC Analysis of an Energy Drink on Ascentis® Express RP-Amide (UV detection)
HPLC Analysis of an Energy Drink on Ascentis® Express RP-Amide (UV detection) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 3 mm
standard
Product No.
Description
Pricing
Caffeine
Sigma Reference Standard, vial of 250 mg
Sorbic acid
99.0-101.0% anhydrous basis
Pyridoxine Hydrochloride (B6)
analytical standard
Nicotinamide (Niacinamide)
analytical standard
Riboflavin (B2)
analytical standard
SS Benzoic Acid
2000 μg/mL in dichloromethane, analytical standard
CONDITIONS
column
Ascentis Express RP-Amide, 5 cm x 3 mm I.D., 2.7 μm particles (53916-U)
mobile phase
[A] water with 0.1% TFA; [B] acetonitrile with 0.1% TFA
flow rate
0.6 mL/min
pressure
1146 psi (79 bar)
column temp.
35 °C
detector
UV, 254 nm
injection
2 μL
sample
Energy Drink Diluted 1:9 in acetonitrile
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of common drink additives used in caffeinated energy drinks.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany