MilliporeSigma
HPLC Analysis of Acidic Drugs on Chromolith® Performance 100- 3 mm
HPLC Analysis of Acidic Drugs on Chromolith® Performance 100- 3 mm application for HPLC
CONDITIONS
column
Chromolith® Performance 100 x 3 mm (1.52001.0001)
mobile phase
[A] acetonitrile; [B] ammoniumacetate 20mM pH=6,8; (28:70, A:B)
flow rate
850 μL/min (u=2mm/s)
pressure
5196 psi (36 bar)
column temp.
ambient
detector
230 nm cell: 1.4 μL
injection
1.0 μL
sample
Indoprofen, Naprofen, Ethacrynic acid, Ibuprofen, Indomethacine
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany