MilliporeSigma
HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide
HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 3 mm
standard
Product No.
Description
Pricing
Atrazine
PESTANAL®, analytical standard
Terbumeton-desethyl
PESTANAL®, analytical standard
Atrazine-desisopropyl
PESTANAL®, analytical standard
Isoproturon
PESTANAL®, analytical standard
Linuron
PESTANAL®, analytical standard
Diuron
PESTANAL®, analytical standard
Chlortoluron
PESTANAL®, analytical standard
Cyanazine
PESTANAL®, analytical standard
Terbumeton
PESTANAL®, analytical standard
Terbuthylazin-desethyl
PESTANAL®, analytical standard
Carbendazim
analytical standard
CONDITIONS
column
Ascentis Express RP-Amide, 15 cm x 3 mm I.D., 2.7 μm particles (53919-U)
mobile phase
[A] water; [B] acetonitrile
gradient
5 to 53% B in 70 min
flow rate
0.4 mL/min
column temp.
35 °C
detector
UV, 240 nm
injection
5 μL
sample
20 mg/L (except terbuthylazin desethyl and terbuthylazine 60 mg/L) in 60:40, water: acetonitrile
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of 16 triazine and phenyl urea compounds. Lower column temperature can improve resolution of the critical pair.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany